Product Details

SNP ID
rs6511701
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:10514391 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGATGCGCGCGTAGAGTGCACAGAT[A/C]GCGGCCAGGATGCCCACGAAGGCGA
Phenotype
MIM: 605146
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
S1PR5 PubMed Links

Gene Details

Gene
S1PR5
Gene Name
sphingosine-1-phosphate receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166215.1 808 Silent Mutation GCG,GCT A207A NP_001159687.1
NM_030760.4 808 Silent Mutation GCG,GCT A207A NP_110387.1

View Full Product Details