Product Details

SNP ID
rs9635390
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90266401 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGAAGCCATGGAGCAGATACGGT[A/G]AGACTCAGGATACCTTGTTTGTATC
Phenotype
MIM: 602293 MIM: 616718
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CIB1 PubMed Links
Additional Information
For this assay, SNP(s) [rs138957600] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CIB1
Gene Name
calcium and integrin binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277764.1 Intron NP_001264693.1
NM_006384.3 Intron NP_006375.2
XM_006720375.1 Intron XP_006720438.1
Gene
LOC400446
Gene Name
PERP, TP53 apoptosis effector pseudogene
There are no transcripts associated with this gene.

Gene
NGRN
Gene Name
neugrin, neurite outgrowth associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033088.1 Intron NP_001028260.2
Gene
TTLL13P
Gene Name
tubulin tyrosine ligase like 13, pseudogene
There are no transcripts associated with this gene.

View Full Product Details