Product Details

SNP ID
rs116709734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48892803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGTCCAGGACACGTGGGTGGG[A/G]GAAGCTGAGCGCTGAGACCAAGGGC
Phenotype
MIM: 603192
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP5G1 PubMed Links

Gene Details

Gene
ATP5G1
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002027.1 Intron NP_001002027.1
NM_005175.2 Intron NP_005166.1
Gene
LOC105371814
Gene Name
uncharacterized LOC105371814
There are no transcripts associated with this gene.

View Full Product Details