Product Details

SNP ID
rs45523834
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.2:233637119 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CCAATGATCTCTTAGGGTTCTCAGA[C/T]GCCATGACTTTCAAGGAGAGAGTAT
Phenotype
MIM: 606435 MIM: 606433
Polymorphism
C/T, Transition substitution
Allele Nomenclature
UGT1A10*1c,g.597T>C UGT1A10*1d,g.597T>C UGT1A10*4c,g.597T>C
Literature Links
UGT1A10 PubMed Links

Gene Details

Gene
UGT1A10
Gene Name
UDP glucuronosyltransferase family 1 member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019075.2 643 Silent Mutation GAC,GAT D199D NP_061948.1
Gene
UGT1A8
Gene Name
UDP glucuronosyltransferase family 1 member A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019076.4 643 Intron NP_061949.3

View Full Product Details