Product Details

SNP ID
hCV25764646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119186078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGCCAGCTTGTACCTGCTTCCC[C/G]TGCCTCCTTGCAGGAAGTTTAAGTT
Phenotype
MIM: 611947 MIM: 607146
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC153 PubMed Links

Gene Details

Gene
CCDC153
Gene Name
coiled-coil domain containing 153
There are no transcripts associated with this gene.

Gene
NLRX1
Gene Name
NLR family member X1
There are no transcripts associated with this gene.

Gene
PDZD3
Gene Name
PDZ domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168468.1 Intron NP_001161940.1
NM_024791.3 Intron NP_079067.3

View Full Product Details