Product Details

SNP ID
rs7856488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122919228 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAGAAAGGGCATAATTGTGGAGA[C/T]GGTTTTGTTCTATTTCACTTACTCT
Phenotype
MIM: 605976
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB26 PubMed Links

Gene Details

Gene
ZBTB26
Gene Name
zinc finger and BTB domain containing 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304363.1 790 Missense Mutation CAT,CGT H236R NP_001291292.1
NM_001304364.1 790 Missense Mutation CAT,CGT H236R NP_001291293.1
NM_020924.3 790 Missense Mutation CAT,CGT H236R NP_065975.1
Gene
ZBTB6
Gene Name
zinc finger and BTB domain containing 6
There are no transcripts associated with this gene.

View Full Product Details