Product Details
- SNP ID
-
rs370616063
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:39636231 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGTGAAAATACTGCAAGAATTCG[A/G]TCTCCGAGAACTTATTTTAATTAGG
- Phenotype
-
MIM: 601081
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs373965264] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCD2
- Gene Name
- ATP binding cassette subfamily D member 2
- Gene
- C12orf40
- Gene Name
- chromosome 12 open reading frame 40
View Full Product Details