Product Details

SNP ID
rs11638723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44956923 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGAAGGGGGAACGATCAGTGACC[C/G]GCGAGCCGCCGACTTCTTGTTCAGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TERB2 PubMed Links

Gene Details

Gene
TERB2
Gene Name
telomere repeat binding bouquet formation protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152448.2 137 Intron NP_689661.1
XM_011521240.2 137 Missense Mutation CCG,CGG P31R XP_011519542.1

View Full Product Details