Product Details
- SNP ID
-
rs61732311
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:23744876 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGCCACATTCTTCACATATGTAC[A/G]ATTTCTCTCCAATATGAATTCTTTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF681
PubMed Links
Gene Details
- Gene
- ZNF681
- Gene Name
- zinc finger protein 681
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138286.2 |
816 |
Missense Mutation |
TCG,TTG |
S225L |
NP_612143.2 |
View Full Product Details