Product Details

SNP ID
rs58974014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:673069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCAACAGTGCTTCAGTTCTCCA[A/G]TGCTCTATGGTCTCACTCAGAGCCC
Phenotype
MIM: 613220
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM18 PubMed Links

Gene Details

Gene
TMEM18
Gene Name
transmembrane protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152834.2 Intron NP_690047.2

View Full Product Details