Product Details

SNP ID
hCV25930808
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.1:182646628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTCCTCACCCCCAGCCTCCCAC[C/T]CCCAATGCCACCGCTTTTGAACACC
Phenotype
MIM: 607189
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RGS8 PubMed Links

Gene Details

Gene
RGS8
Gene Name
regulator of G-protein signaling 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102450.2 812 UTR 3 NP_001095920.1
NM_033345.3 812 UTR 3 NP_203131.1

View Full Product Details