Product Details

SNP ID
rs201126475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:83018050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTATGGTATTATTTAGCCAAGA[T/A]GTATTATAATGCTAAATTATGTATA
Phenotype
MIM: 608530
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
BTBD1 PubMed Links

Gene Details

Gene
BTBD1
Gene Name
BTB domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011885.1 1669 UTR 3 NP_001011885.1
NM_025238.3 1669 UTR 3 NP_079514.1
Gene
C15orf40
Gene Name
chromosome 15 open reading frame 40
There are no transcripts associated with this gene.

View Full Product Details