Product Details

SNP ID
rs61738562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:82989048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACACTGCCGAAGCTGTTCCAAA[G/T]TTTGAAGAGATGTTTGCTAGTAGAT
Phenotype
MIM: 614547 MIM: 604799
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C15orf40 PubMed Links

Gene Details

Gene
C15orf40
Gene Name
chromosome 15 open reading frame 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160113.1 510 Intron NP_001153585.1
NM_001160114.1 510 Intron NP_001153586.1
NM_001160115.1 510 Intron NP_001153587.1
NM_001160116.1 510 UTR 3 NP_001153588.1
NM_144597.2 510 Intron NP_653198.2
XM_006720385.2 510 Intron XP_006720448.1
XM_011521212.2 510 Intron XP_011519514.1
XM_011521213.2 510 Intron XP_011519515.1
XM_011521214.2 510 Intron XP_011519516.1
Gene
FAM103A1
Gene Name
family with sequence similarity 103 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031452.3 510 Intron NP_113640.1
Gene
HOMER2
Gene Name
homer scaffolding protein 2
There are no transcripts associated with this gene.

View Full Product Details