Product Details

SNP ID
rs34257411
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:114674038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGTTACTTAGTGGTGACATGGC[A/G]ATGGGAATTTGGGCTAAGAAAAACA
Phenotype
MIM: 102770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMPD1 PubMed Links

Gene Details

Gene
AMPD1
Gene Name
adenosine monophosphate deaminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000036.2 1992 Silent Mutation ATC,ATT I648I NP_000027.2
NM_001172626.1 1992 Silent Mutation ATC,ATT I644I NP_001166097.1
Gene
DENND2C
Gene Name
DENN domain containing 2C
There are no transcripts associated with this gene.

View Full Product Details