Product Details
- SNP ID
-
rs17115997
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:113801625 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTCATCACATTAATGCCCTCAGT[A/T]ACGGAGTGATCATCATTTGTTTCAA
- Phenotype
-
MIM: 610748
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
USP28
PubMed Links
Gene Details
- Gene
- USP28
- Gene Name
- ubiquitin specific peptidase 28
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001301029.1 |
3170 |
Silent Mutation |
GTA,GTT |
V815V |
NP_001287958.1 |
NM_020886.3 |
3170 |
Silent Mutation |
GTA,GTT |
V972V |
NP_065937.1 |
XM_005271630.2 |
3170 |
Silent Mutation |
GTA,GTT |
V1035V |
XP_005271687.1 |
XM_005271631.2 |
3170 |
Silent Mutation |
GTA,GTT |
V1034V |
XP_005271688.1 |
XM_005271632.2 |
3170 |
Silent Mutation |
GTA,GTT |
V1032V |
XP_005271689.1 |
XM_005271633.4 |
3170 |
Silent Mutation |
GTA,GTT |
V1013V |
XP_005271690.1 |
XM_005271636.2 |
3170 |
Silent Mutation |
GTA,GTT |
V975V |
XP_005271693.1 |
XM_005271637.2 |
3170 |
Silent Mutation |
GTA,GTT |
V973V |
XP_005271694.1 |
XM_005271638.2 |
3170 |
Silent Mutation |
GTA,GTT |
V941V |
XP_005271695.1 |
XM_005271639.2 |
3170 |
Silent Mutation |
GTA,GTT |
V940V |
XP_005271696.1 |
XM_011542936.2 |
3170 |
Silent Mutation |
GTA,GTT |
V1009V |
XP_011541238.1 |
XM_011542938.1 |
3170 |
Silent Mutation |
GTA,GTT |
V910V |
XP_011541240.1 |
XM_011542941.1 |
3170 |
Silent Mutation |
GTA,GTT |
V682V |
XP_011541243.1 |
XM_011542942.1 |
3170 |
Silent Mutation |
GTA,GTT |
V591V |
XP_011541244.1 |
XM_017018056.1 |
3170 |
Silent Mutation |
GTA,GTT |
V1008V |
XP_016873545.1 |
XM_017018057.1 |
3170 |
Silent Mutation |
GTA,GTT |
V969V |
XP_016873546.1 |
XM_017018058.1 |
3170 |
Silent Mutation |
GTA,GTT |
V946V |
XP_016873547.1 |
XM_017018059.1 |
3170 |
Silent Mutation |
GTA,GTT |
V914V |
XP_016873548.1 |
XM_017018060.1 |
3170 |
Silent Mutation |
GTA,GTT |
V909V |
XP_016873549.1 |
XM_017018061.1 |
3170 |
Silent Mutation |
GTA,GTT |
V848V |
XP_016873550.1 |
XM_017018062.1 |
3170 |
Silent Mutation |
GTA,GTT |
V847V |
XP_016873551.1 |
XM_017018063.1 |
3170 |
Silent Mutation |
GTA,GTT |
V747V |
XP_016873552.1 |
XM_017018064.1 |
3170 |
Silent Mutation |
GTA,GTT |
V685V |
XP_016873553.1 |
XM_017018065.1 |
3170 |
Silent Mutation |
GTA,GTT |
V620V |
XP_016873554.1 |
XM_017018066.1 |
3170 |
Silent Mutation |
GTA,GTT |
V620V |
XP_016873555.1 |
XM_017018067.1 |
3170 |
Silent Mutation |
GTA,GTT |
V620V |
XP_016873556.1 |
XM_017018068.1 |
3170 |
Silent Mutation |
GTA,GTT |
V588V |
XP_016873557.1 |
XM_017018069.1 |
3170 |
Intron |
|
|
XP_016873558.1 |
View Full Product Details