Product Details

SNP ID
rs75850792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40740865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAGAACATTCAGAGCCAATGTC[A/G]TAGAGCTTTTTATTTTGCATCTGGA
Phenotype
MIM: 606476 MIM: 182285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf54 PubMed Links

Gene Details

Gene
C19orf54
Gene Name
chromosome 19 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198476.3 2284 UTR 3 NP_940878.3
XM_005258776.4 2284 UTR 3 XP_005258833.1
XM_005258778.4 2284 UTR 3 XP_005258835.1
XM_006723152.2 2284 Intron XP_006723215.1
XM_006723153.2 2284 Intron XP_006723216.1
XM_011526774.2 2284 UTR 3 XP_011525076.1
XM_011526775.1 2284 Intron XP_011525077.1
XM_011526776.2 2284 UTR 3 XP_011525078.1
XM_011526777.2 2284 UTR 3 XP_011525079.1
XM_011526778.2 2284 UTR 3 XP_011525080.1
XM_011526779.2 2284 UTR 3 XP_011525081.1
XM_011526782.2 2284 Intron XP_011525084.1
XM_011526783.2 2284 UTR 3 XP_011525085.1
XM_011526784.2 2284 UTR 3 XP_011525086.1
XM_011526785.2 2284 UTR 3 XP_011525087.1
XM_011526786.2 2284 Intron XP_011525088.1
XM_017026620.1 2284 UTR 3 XP_016882109.1
XM_017026621.1 2284 UTR 3 XP_016882110.1
XM_017026622.1 2284 UTR 3 XP_016882111.1
XM_017026623.1 2284 UTR 3 XP_016882112.1
XM_017026624.1 2284 UTR 3 XP_016882113.1
XM_017026625.1 2284 Intron XP_016882114.1
XM_017026626.1 2284 UTR 3 XP_016882115.1
XM_017026627.1 2284 UTR 3 XP_016882116.1
Gene
ITPKC
Gene Name
inositol-trisphosphate 3-kinase C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025194.2 2284 Intron NP_079470.1
XM_017027324.1 2284 Intron XP_016882813.1
Gene
SNRPA
Gene Name
small nuclear ribonucleoprotein polypeptide A
There are no transcripts associated with this gene.

View Full Product Details