Product Details

SNP ID
rs149431964
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:6634766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAGGACGCTGGACCTGCAGGAG[C/T]GGGGAGCTGCAGTGCCACCTGCTGG
Phenotype
MIM: 614827 MIM: 612532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC11 PubMed Links

Gene Details

Gene
DNAJC11
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018198.3 1212 UTR 3 NP_060668.2
XM_017001745.1 1212 UTR 3 XP_016857234.1
Gene
THAP3
Gene Name
THAP domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195752.1 1212 Intron NP_001182681.1
NM_001195753.1 1212 Intron NP_001182682.1
NM_138350.3 1212 UTR 3 NP_612359.2
XM_005263532.3 1212 Intron XP_005263589.1
XM_011542400.2 1212 Intron XP_011540702.1
XM_011542401.2 1212 Intron XP_011540703.1
XM_017002761.1 1212 UTR 3 XP_016858250.1

View Full Product Details