Product Details

SNP ID
rs117567558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144532459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCTGCGGCTTCATGATTCCGCC[G/T]GGACACTCCCTCCGGTAGGCCCCTG
Phenotype
MIM: 615880
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP39 PubMed Links

Gene Details

Gene
ARHGAP39
Gene Name
Rho GTPase activating protein 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308207.1 Intron NP_001295136.1
NM_001308208.1 Intron NP_001295137.1
NM_025251.2 Intron NP_079527.1
XM_011517308.1 Intron XP_011515610.1
XM_011517309.1 Intron XP_011515611.1
XM_011517312.2 Intron XP_011515614.1
XM_017013870.1 Intron XP_016869359.1
XM_017013871.1 Intron XP_016869360.1
Gene
C8orf82
Gene Name
chromosome 8 open reading frame 82
There are no transcripts associated with this gene.

Gene
LRRC14
Gene Name
leucine rich repeat containing 14
There are no transcripts associated with this gene.

Gene
LRRC24
Gene Name
leucine rich repeat containing 24
There are no transcripts associated with this gene.

View Full Product Details