Product Details
- SNP ID
-
rs34353835
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:73985325 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCCCAGGGCTCCTCACCTGGCCC[C/T]GGGGACGGGGCACCTCCTGCCATTG
- Phenotype
-
MIM: 102578
MIM: 608326
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PML
PubMed Links
Gene Details
- Gene
- PML
- Gene Name
- promyelocytic leukemia
There are no transcripts associated with this gene.
- Gene
- STOML1
- Gene Name
- stomatin like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256672.1 |
1813 |
Silent Mutation |
CCA,CCG |
P261P |
NP_001243601.1 |
NM_001256673.1 |
1813 |
Silent Mutation |
CCA,CCG |
P211P |
NP_001243602.1 |
NM_001256674.1 |
1813 |
Silent Mutation |
CCA,CCG |
P211P |
NP_001243603.1 |
NM_001256675.1 |
1813 |
Silent Mutation |
CCA,CCG |
P261P |
NP_001243604.1 |
NM_001256676.1 |
1813 |
Silent Mutation |
CCA,CCG |
P174P |
NP_001243605.1 |
NM_001256677.1 |
1813 |
Silent Mutation |
CCA,CCG |
P219P |
NP_001243606.1 |
NM_001324226.1 |
1813 |
Silent Mutation |
CCA,CCG |
P105P |
NP_001311155.1 |
NM_001324227.1 |
1813 |
Silent Mutation |
CCA,CCG |
P174P |
NP_001311156.1 |
NM_001324228.1 |
1813 |
Silent Mutation |
CCA,CCG |
P174P |
NP_001311157.1 |
NM_001324229.1 |
1813 |
Silent Mutation |
CCA,CCG |
P105P |
NP_001311158.1 |
NM_001324230.1 |
1813 |
Silent Mutation |
CCA,CCG |
P219P |
NP_001311159.1 |
NM_004809.4 |
1813 |
Silent Mutation |
CCA,CCG |
P261P |
NP_004800.2 |
XM_005254792.4 |
1813 |
Silent Mutation |
CCA,CCG |
P219P |
XP_005254849.1 |
XM_006720773.3 |
1813 |
Silent Mutation |
CCA,CCG |
P219P |
XP_006720836.1 |
View Full Product Details