Product Details

SNP ID
rs8112713
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15770941 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTATTTCATCTGACATTTGACAT[A/C]AAGCTCCATTCTATCACTGTAATTC
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CYP4F24P PubMed Links

Gene Details

Gene
CYP4F24P
Gene Name
cytochrome P450 family 4 subfamily F member 24, pseudogene
There are no transcripts associated with this gene.

View Full Product Details