Product Details

SNP ID
rs78328553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:97096725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCAAATTTATATTTGGAAAAATC[C/T]GAATAAAGGAAATCTCCTGGGAAGA
Phenotype
MIM: 610466
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIX1 PubMed Links

Gene Details

Gene
LIX1
Gene Name
limb and CNS expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153234.4 Intron NP_694966.3

View Full Product Details