Product Details

SNP ID
rs12251842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69180310 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCTCGATGTCCTTCTCCCGTGCC[C/T]TATTGTGGGCTCGGCTCCCGGCGGG
Phenotype
MIM: 605122 MIM: 605506
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUPV3L1 PubMed Links

Gene Details

Gene
SUPV3L1
Gene Name
Suv3 like RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301683.1 107 UTR 5 NP_001288612.1
NM_001323584.1 107 UTR 5 NP_001310513.1
NM_001323585.1 107 UTR 5 NP_001310514.1
NM_001323586.1 107 UTR 5 NP_001310515.1
NM_001323587.1 107 UTR 5 NP_001310516.1
NM_001323588.1 107 UTR 5 NP_001310517.1
NM_003171.4 107 Silent Mutation CTA,TTA L7L NP_003162.2
Gene
VPS26A
Gene Name
VPS26, retromer complex component A
There are no transcripts associated with this gene.

View Full Product Details