Product Details

SNP ID
rs34666550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18392051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGGCCCAGTGACTGCAGGTTA[C/T]AGTAGACAGGCTGGGAAGCCAGGTC
Phenotype
MIM: 605312 MIM: 607518
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GDF15 PubMed Links

Gene Details

Gene
GDF15
Gene Name
growth differentiation factor 15
There are no transcripts associated with this gene.

Gene
LRRC25
Gene Name
leucine rich repeat containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145256.2 1050 Missense Mutation TAT,TGT Y285C NP_660299.2
XM_005259739.3 1050 Missense Mutation TAT,TGT Y285C XP_005259796.1
Gene
MIR3189
Gene Name
microRNA 3189
There are no transcripts associated with this gene.

View Full Product Details