Product Details

SNP ID
rs3739950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136791795 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTCACAGGTAGGCTGCGTCCAGGT[A/G]TCCTGCGGCTGGGAGAGAACGCACC
Phenotype
MIM: 615955
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC183 PubMed Links

Gene Details

Gene
CCDC183
Gene Name
coiled-coil domain containing 183
There are no transcripts associated with this gene.

Gene
TMEM141
Gene Name
transmembrane protein 141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032928.3 Intron NP_116317.1

View Full Product Details