Product Details

SNP ID
hCV25997897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48923029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAGAGAGTGCAGGGGTGGGCTA[G/C]CCAGAGGGAAGTGAGGAGGAGGGAA
Phenotype
MIM: 300713 MIM: 300295
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
OTUD5 PubMed Links

Gene Details

Gene
OTUD5
Gene Name
OTU deubiquitinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136157.1 1907 UTR 3 NP_001129629.1
NM_001136158.1 1907 Intron NP_001129630.1
NM_001136159.1 1907 UTR 3 NP_001129631.1
NM_017602.3 1907 UTR 3 NP_060072.1
Gene
PIM2
Gene Name
Pim-2 proto-oncogene, serine/threonine kinase
There are no transcripts associated with this gene.

View Full Product Details