Product Details
- SNP ID
-
rs10117333
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:136207605 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCAAATGGTAAAAGACATCATGAC[A/C]AAAGCTTGGGAGATCAGTGCCTGTG
- Phenotype
-
MIM: 600577
MIM: 612860
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LHX3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79321298] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LHX3
- Gene Name
- LIM homeobox 3
There are no transcripts associated with this gene.
- Gene
- QSOX2
- Gene Name
- quiescin sulfhydryl oxidase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181701.3 |
3258 |
UTR 3 |
|
|
NP_859052.3 |
View Full Product Details