Product Details
- SNP ID
-
rs12030054
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
37
- Location
-
Chr.1:230979608 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGCTACTTCCTTCTAGCTCTTCT[C/T]CCTTTGACAGAGAGATGGTACCATC
- Phenotype
-
MIM: 611647
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARV1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78129525] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARV1
- Gene Name
- ARV1 homolog, fatty acid homeostasis modulator
- Gene
- TTC13
- Gene Name
- tetratricopeptide repeat domain 13
View Full Product Details