Product Details
- SNP ID
-
rs9835850
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:184567867 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATAGGTAGGCTGGAAGCGTGGGTC[C/T]GCCTAAGGCAAGCCAGATCTCTGTG
- Phenotype
-
MIM: 601839
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EPHB3
PubMed Links
Gene Details
- Gene
- EPHB3
- Gene Name
- EPH receptor B3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004443.3 |
|
Intron |
|
|
NP_004434.2 |
View Full Product Details