Product Details
- SNP ID
-
hCV26116894
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:51910145 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGTCTGTCAGACTGTAAGCCCTG[G/T]AGGCTTAACTATTCTGCCTTATGAC
- Phenotype
-
MIM: 179510
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RAB3B
PubMed Links
Gene Details
- Gene
- RAB3B
- Gene Name
- RAB3B, member RAS oncogene family
There are no transcripts associated with this gene.
View Full Product Details