Product Details

SNP ID
rs6684297
Assay Type
Functionally tested
NCBI dbSNP Submissions
64
Location
Chr.1:182378532 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTAAGAACAATTATACCTTCTAC[A/G]TCTTACCACTCCCCATTCCTAGTGA
Phenotype
MIM: 138290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLUL PubMed Links

Gene Details

Gene
GLUL
Gene Name
glutamate-ammonia ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033044.3 7892 UTR 3 NP_001028216.1
NM_001033056.3 7892 UTR 3 NP_001028228.1
NM_002065.6 7892 UTR 3 NP_002056.2
XM_006711278.1 7892 Intron XP_006711341.1

View Full Product Details