Product Details

SNP ID
rs4414843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52050162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCTTCAAGAAGGGCCCGCTGA[A/G]GAAACAGAGGCTTCACACATGTGCA
Phenotype
MIM: 602749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP7 PubMed Links

Gene Details

Gene
DUSP7
Gene Name
dual specificity phosphatase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001947.3 1992 UTR 3 NP_001938.2

View Full Product Details