Product Details

SNP ID
rs1802462
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75038949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAATTTTATAAATTCTCAATTG[T/G]TTGGTGAGGCCAATAGGGATACTTT
Phenotype
MIM: 613422
Polymorphism
T/G, Transversion Substitution
Allele Nomenclature
Literature Links
ATP5H PubMed Links
Additional Information
For this assay, SNP(s) [rs6968] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ATP5H
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003785.1 Intron NP_001003785.1
NM_006356.2 Intron NP_006347.1
Gene
KCTD2
Gene Name
potassium channel tetramerization domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015353.2 Intron NP_056168.1
XM_011524575.2 Intron XP_011522877.1

View Full Product Details