Product Details

SNP ID
hCV26242680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41586219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCTGGGGTGGACTCTGTCCTAT[A/G]GAGAAATCTTAAGGTCTCAGGGGCC
Phenotype
MIM: 148066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT14 PubMed Links

Gene Details

Gene
KRT14
Gene Name
keratin 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000526.4 Intron NP_000517.2

View Full Product Details