Product Details
- SNP ID
-
rs72559714
- Assay Type
- DME
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:17394337 on Build GRCh38
- Set Membership
-
DME
Validated
Inventoried
- Context Sequence [VIC/FAM]
- CTGGTCTTCCTCACGAAGGCCCGGG[C/T]CAGGCAGAACAGCTGCCTCTGTCCC
- Phenotype
-
MIM: 600509
MIM: 600937
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ABCC8
PubMed Links
Gene Details
- Gene
- ABCC8
- Gene Name
- ATP binding cassette subfamily C member 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000352.4 |
4603 |
Missense Mutation |
ACC,GCC |
T1492A |
NP_000343.2 |
NM_001287174.1 |
4603 |
Missense Mutation |
ACC,GCC |
T1493A |
NP_001274103.1 |
XM_011520331.2 |
4603 |
Missense Mutation |
ACC,GCC |
T1492A |
XP_011518633.1 |
XM_017018197.1 |
4603 |
Missense Mutation |
ACC,GCC |
T1515A |
XP_016873686.1 |
XM_017018198.1 |
4603 |
Missense Mutation |
ACC,GCC |
T1514A |
XP_016873687.1 |
XM_017018199.1 |
4603 |
Missense Mutation |
ACC,GCC |
T1514A |
XP_016873688.1 |
XM_017018200.1 |
4603 |
Missense Mutation |
ACC,GCC |
T1491A |
XP_016873689.1 |
XM_017018201.1 |
4603 |
Missense Mutation |
GAC,GGC |
D1480G |
XP_016873690.1 |
XM_017018202.1 |
4603 |
Missense Mutation |
ACC,GCC |
T1014A |
XP_016873691.1 |
XM_017018203.1 |
4603 |
Intron |
|
|
XP_016873692.1 |
XM_017018204.1 |
4603 |
Missense Mutation |
ACC,GCC |
T811A |
XP_016873693.1 |
- Gene
- KCNJ11
- Gene Name
- potassium voltage-gated channel subfamily J member 11
There are no transcripts associated with this gene.
View Full Product Details