Product Details

SNP ID
rs4458216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:110743558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACAAACCCAAGTTCAGAGACTC[A/G]GGGAGGAGGGCCGGGTGGAGGTCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACOXL PubMed Links

Gene Details

Gene
ACOXL
Gene Name
acyl-CoA oxidase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142807.1 Intron NP_001136279.1
XM_011511404.2 Intron XP_011509706.1
XM_011511405.2 Intron XP_011509707.1
XM_011511406.2 Intron XP_011509708.1
XM_011511407.2 Intron XP_011509709.1
XM_011511409.1 Intron XP_011509711.1
XM_011511411.2 Intron XP_011509713.1
XM_011511413.1 Intron XP_011509715.1
XM_011511414.2 Intron XP_011509716.1
XM_011511415.2 Intron XP_011509717.1
XM_011511416.2 Intron XP_011509718.1
XM_011511419.2 Intron XP_011509721.1
XM_011511420.2 Intron XP_011509722.1
XM_011511421.2 Intron XP_011509723.1
XM_011511422.1 Intron XP_011509724.1
XM_011511423.1 Intron XP_011509725.1
XM_011511427.2 Intron XP_011509729.1
XM_011511428.2 Intron XP_011509730.1
XM_011511429.2 Intron XP_011509731.1
XM_011511432.2 Intron XP_011509734.1
XM_011511434.2 Intron XP_011509736.1
XM_017004431.1 Intron XP_016859920.1
XM_017004432.1 Intron XP_016859921.1
XM_017004433.1 Intron XP_016859922.1
XM_017004434.1 Intron XP_016859923.1

View Full Product Details