Product Details

SNP ID
rs6821591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:23795377 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTTTGAAGTTCTAGGTTTTAAG[C/T]GTGTCTTCATGGAACTGCTGCCATT
Phenotype
MIM: 604517
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPARGC1A PubMed Links

Gene Details

Gene
PPARGC1A
Gene Name
PPARG coactivator 1 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013261.3 2677 UTR 3 NP_037393.1
XM_005248131.4 2677 UTR 3 XP_005248188.1
XM_005248132.1 2677 UTR 3 XP_005248189.1
XM_005248134.4 2677 UTR 3 XP_005248191.1
XM_011513765.2 2677 UTR 3 XP_011512067.1
XM_011513766.1 2677 UTR 3 XP_011512068.1
XM_011513767.2 2677 UTR 3 XP_011512069.1
XM_011513768.1 2677 UTR 3 XP_011512070.1
XM_011513769.2 2677 Intron XP_011512071.1
XM_011513770.2 2677 UTR 3 XP_011512072.1
XM_011513771.1 2677 UTR 3 XP_011512073.1
XM_017007664.1 2677 UTR 3 XP_016863153.1

View Full Product Details