Product Details

SNP ID
rs9912101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47469712 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTAAGTTTTTTCCATCGTGAAAG[C/T]GTAAATGAAATTAAGAAAAAAACAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL45P2 PubMed Links

Gene Details

Gene
MRPL45P2
Gene Name
mitochondrial ribosomal protein L45 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details