Product Details

SNP ID
rs238193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49278434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGACATGGCGAGGAAGCGGACA[A/G]GCCCGGGTGGCCCGGCGTGTAGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNORD12 PubMed Links

Gene Details

Gene
SNORD12
Gene Name
small nucleolar RNA, C/D box 12
There are no transcripts associated with this gene.

Gene
SNORD12B
Gene Name
small nucleolar RNA, C/D box 12B
There are no transcripts associated with this gene.

Gene
SNORD12C
Gene Name
small nucleolar RNA, C/D box 12C
There are no transcripts associated with this gene.

Gene
ZFAS1
Gene Name
ZNFX1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNFX1
Gene Name
zinc finger NFX1-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021035.2 Intron NP_066363.1
XM_006723836.1 Intron XP_006723899.1
XM_011528924.1 Intron XP_011527226.1
XM_017027971.1 Intron XP_016883460.1

View Full Product Details