Product Details

SNP ID
rs2322030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:165262168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTGTAGTGTTGGATATAGTATT[C/T]ATAGTGAAGATAGTGGTTCAATCCC
Phenotype
MIM: 605774
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL2 PubMed Links

Gene Details

Gene
KLHL2
Gene Name
kelch like family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161521.1 Intron NP_001154993.1
NM_001161522.1 Intron NP_001154994.1
NM_007246.3 Intron NP_009177.3
XM_011531572.2 Intron XP_011529874.1
XM_011531575.2 Intron XP_011529877.1
XM_011531576.2 Intron XP_011529878.1
XM_017007674.1 Intron XP_016863163.1
XM_017007675.1 Intron XP_016863164.1
XM_017007676.1 Intron XP_016863165.1
XM_017007677.1 Intron XP_016863166.1

View Full Product Details