Product Details
- SNP ID
-
hCV26811098
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:70251453 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCAAATTTGGTTTCACAAATATG[T/C]AGTAAATTTAAAAATGTAAAACATG
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SMA4
PubMed Links
Gene Details
- Gene
- SMA4
- Gene Name
- glucuronidase beta pseudogene
There are no transcripts associated with this gene.
View Full Product Details