Product Details

SNP ID
rs401933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38479544 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGCTAGGAGCGTAGTAAATAGGC[C/T]GGGAAATGACTTCATAGATTCGATT
Phenotype
MIM: 608469
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX17 PubMed Links

Gene Details

Gene
DDX17
Gene Name
DEAD-box helicase 17
There are no transcripts associated with this gene.

Gene
KDELR3
Gene Name
KDEL endoplasmic reticulum protein retention receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006855.3 Intron NP_006846.1
NM_016657.2 Intron NP_057839.1

View Full Product Details