Product Details

SNP ID
rs11237244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77624291 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAATAATAGTAATAATAAAACA[A/G]TATAAATGATCCAGGTTCTCAAGGT
Phenotype
MIM: 602158
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLNS1A PubMed Links

Gene Details

Gene
CLNS1A
Gene Name
chloride nucleotide-sensitive channel 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293.2 Intron NP_001284.1
NM_001311199.1 Intron NP_001298128.1
NM_001311200.1 Intron NP_001298129.1
NM_001311201.1 Intron NP_001298130.1
NM_001311202.1 Intron NP_001298131.1
Gene
LOC107987163
Gene Name
uncharacterized LOC107987163
There are no transcripts associated with this gene.

View Full Product Details