Product Details
- SNP ID
-
hCV26953307
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:5315204 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCTGGGGTCCCCAGCTTGGAGCT[C/T]CCAAGACCCTGGTCTGTCCCGGGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TNRC18
PubMed Links
Gene Details
- Gene
- TNRC18
- Gene Name
- trinucleotide repeat containing 18
View Full Product Details