Product Details

SNP ID
rs4073310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137077767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCGGCCCCTGCCCCCAGAGCGC[G/T]TGGGCAGGGCCAGCCGCAGCGACCC
Phenotype
MIM: 604346 MIM: 612057
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAN1B1 PubMed Links

Gene Details

Gene
MAN1B1
Gene Name
mannosidase alpha class 1B member 1
There are no transcripts associated with this gene.

Gene
MAN1B1-AS1
Gene Name
MAN1B1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
SAPCD2
Gene Name
suppressor APC domain containing 2
There are no transcripts associated with this gene.

Gene
UAP1L1
Gene Name
UDP-N-acetylglucosamine pyrophosphorylase 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207309.2 293 Missense Mutation GTG,TTG V79L NP_997192.2
XM_006717317.3 293 Missense Mutation GTG,TTG V79L XP_006717380.1
XM_011519182.2 293 Intron XP_011517484.1

View Full Product Details