Product Details
- SNP ID
-
rs1818396
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:131603018 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGATTTGCGCGGCAAAAAGAGTCAT[A/G]TAAGATGGTCTATGAGATGACTTTG
- Phenotype
-
MIM: 611266
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
POTEI
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142540546,rs148426080] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POTEI
- Gene Name
- POTE ankyrin domain family member I
- Gene
- POTEKP
- Gene Name
- POTE ankyrin domain family member K, pseudogene
There are no transcripts associated with this gene.
View Full Product Details