Product Details

SNP ID
rs11880523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48033606 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTACCTTCCAGGAACATAACTAGGA[A/G]TTAGCCAGGGGAGAGGGGAAAGGGT
Phenotype
MIM: 607315 MIM: 607443
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CABP5 PubMed Links

Gene Details

Gene
CABP5
Gene Name
calcium binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019855.4 Intron NP_062829.1
XM_017026992.1 Intron XP_016882481.1
Gene
ELSPBP1
Gene Name
epididymal sperm binding protein 1
There are no transcripts associated with this gene.

View Full Product Details