Product Details
- SNP ID
-
rs4301502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:73153267 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAGGTCTGCAAAGGTTGATTTCT[A/G]AATATGACGTTAGCTCTCTCTAGAC
- Phenotype
-
MIM: 151690
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA1
PubMed Links
Gene Details
- Gene
- ANXA1
- Gene Name
- annexin A1
View Full Product Details