Product Details

SNP ID
rs6853435
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:153784864 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACACACCAAAAGGTACCTGCCCTT[A/G]GAAAAGGATAAAAGCAAGAATGCCT
Phenotype
MIM: 604157
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SFRP2 PubMed Links

Gene Details

Gene
SFRP2
Gene Name
secreted frizzled related protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003013.2 Intron NP_003004.1

View Full Product Details