Product Details
- SNP ID
-
rs9444346
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:85455913 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACAGTAGAAAAAGCAGTTTGTGTGT[A/G]TGTGTGTGTTTGTGCTTTTGTATGC
- Phenotype
-
MIM: 129190
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NT5E
PubMed Links
Gene Details
- Gene
- NT5E
- Gene Name
- 5'-nucleotidase ecto
View Full Product Details