Product Details

SNP ID
rs4919690
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102856743 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTAGAAATCTCAGATGGTATGTTA[C/T]GTGTCTGGTCTGTTTATGTAGGAAA
Phenotype
MIM: 616600
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
BORCS7 PubMed Links

Gene Details

Gene
BORCS7
Gene Name
BLOC-1 related complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136200.1 Intron NP_001129672.1
NM_144591.3 Intron NP_653192.2
Gene
BORCS7-ASMT
Gene Name
BORCS7-ASMT readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details